EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Uptake of perfluorooctanoic acid by Caco-2 cells: Involvement of organic anion transporting polypeptides.
PMID 28552774
Abstract
The mechanism underlying the intestinal absorption of perfluorooctanoic acid (PFOA) was investigated using Caco-2 cells. The uptake of PFOA from the apical membrane of Caco-2 cells was fast, and pH, temperature, and concentration dependent, but Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B17905
Benzyl bromide, reagent grade, 98%
C7H7Br