EMAIL THIS PAGE TO A FRIEND
Food chemistry
Graphene oxide-sensitized molecularly imprinted opto-polymers for charge-transfer fluorescent sensing of cyanoguanidine.
PMID 28554618
Abstract
The hierarchical structuring of materials offers exciting opportunities to construct functional sensors. Multiple processes were combined to create complex materials for the selective detection of cyanoguanidine (CYA) using graphene oxide-sensitized molecularly imprinted opto-polymers (MIOP). Molecular imprinting was used to construct molecular-scale analyte-selective cavities, graphene oxide was introduced to provide a platform for the polymerization, and increase the stability and binding kinetic properties, and 3-methacryloxy propyl trimethoxy silane-modified quantum dots were combined with a functional monomer to increase the fluorescence quantum yield. Polymer cross-linking and fluorescence intensity were optimized for molecular recognition and opto-sensing detection. Selective and sensitive, fluorescence sensing of CYA was possible at concentrations as low as to 1.6μM. It could be applied to the rapid and cost-effective monitoring of CYA in infant formula. The approach is generic and applicable to many molecules and conventional opto-sensors, based on molecularly imprinted polymer formulations, individually or in multiplexed arrays.
Related Materials