EMAIL THIS PAGE TO A FRIEND
Nature communications
JunB is essential for IL-23-dependent pathogenicity of Th17 cells.
PMID 28555647
Abstract
CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0447
SIS3, ≥98% (HPLC), powder
C28H27N3O3 · HCl