EMAIL THIS PAGE TO A FRIEND
Epilepsia
Cacna1g is a genetic modifier of epilepsy in a mouse model of Dravet syndrome.
PMID 28556246
Abstract
Dravet syndrome, an early onset epileptic encephalopathy, is most often caused by de novo mutation of the neuronal voltage-gated sodium channel gene SCN1A. Mouse models with deletion of Scn1a recapitulate Dravet syndrome phenotypes, including spontaneous generalized tonic-clonic seizures, susceptibility to seizures induced by elevated body temperature, and elevated risk of sudden unexpected death in epilepsy. Importantly, the epilepsy phenotype of Dravet mouse models is highly strain-dependent, suggesting a strong influence of genetic modifiers. We previously identified Cacna1g, encoding the Cav3.1 subunit of the T-type calcium channel family, as an epilepsy modifier in the Scn2a