EMAIL THIS PAGE TO A FRIEND
Plant, cell & environment
SUV2, which encodes an ATR-related cell cycle checkpoint and putative plant ATRIP, is required for aluminium-dependent root growth inhibition in Arabidopsis.
PMID 28556304
Abstract
A suppressor mutagenesis screen was conducted in order to identify second site mutations that could reverse the extreme hypersensitivity to aluminium (Al) seen for the Arabidopsis mutant, als3-1. From this screen, it was found that a loss-of-function mutation in the previously described SUV2 (SENSITIVE TO UV 2), which encodes a putative plant ATRIP homologue that is a component of the ATR-dependent cell checkpoint response, reversed the als3-1 phenotype. This included prevention of hallmarks associated with als3-1 including Al-dependent terminal differentiation of the root tip and transition to endoreduplication. From this analysis, SUV2 was determined to be required for halting cell cycle progression and triggering loss of the quiescent centre (QC) following exposure to Al. In conjunction with this, SUV2 was found to have a similar role as ATR, ALT2 and SOG1 in Al-dependent stoppage of root growth, all of which are required for promotion of expression of a suite of genes that likely are part of an Al-dependent DNA damage transcriptional response. This work argues that these Al response factors work together to detect Al-dependent damage and subsequently activate a DNA damage response pathway that halts the cell cycle and subsequently promotes QC differentiation and entrance into endocycling.