EMAIL THIS PAGE TO A FRIEND
ACS synthetic biology
Engineered Assimilation of Exogenous and Endogenous Formate in Escherichia coli.
PMID 28558223
Abstract
Decoupling biorefineries from land use and agriculture is a major challenge. As formate can be produced from various sources, e.g., electrochemical reduction of CO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
605409
D-Glucose-3-13C, 99 atom % 13C, 99% (CP)
13CC5H12O6
I4125
Inosine, ≥99% (HPLC)
C10H12N4O5