EMAIL THIS PAGE TO A FRIEND
Chinese journal of natural medicines
Triptolide reduces prostate size and androgen level on testosterone-induced benign prostatic hyperplasia in Sprague Dawley rats.
PMID 28558869
Abstract
Benign prostatic hyperplasia (BPH) is an age-related disease of unknown etiology, characterized by prostatic enlargement coincident with distinct alterations in tissue histology. In the present study, we investigated whether triptolide can prevent testosterone-induced prostatic hyperplasia in rats. Castration was performed via the scrotal route after urethane aesthesia. BPH was induced in experimental groups by daily subcutaneous injections of testosterone propionate (TP) for two weeks. Triptolide was administered daily by oral gavage at a dose of 100 and 50 μg·kg
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MHS16
Hematoxylin Solution, Mayer’s