EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
AraC-like transcriptional activator CuxR binds c-di-GMP by a PilZ-like mechanism to regulate extracellular polysaccharide production.
PMID 28559336
Abstract
Cyclic dimeric GMP (c-di-GMP) has emerged as a key regulatory player in the transition between planktonic and sedentary biofilm-associated bacterial lifestyles. It controls a multitude of processes including production of extracellular polysaccharides (EPSs). The PilZ domain, consisting of an N-terminal "RxxxR" motif and a β-barrel domain, represents a prototype c-di-GMP receptor. We identified a class of c-di-GMP-responsive proteins, represented by the AraC-like transcription factor CuxR in plant symbiotic α-proteobacteria. In
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6508
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
C4H8N2O4