EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
APMAP interacts with lysyl oxidase-like proteins, and disruption of
PMID 28559441
Abstract
Adipocyte plasma membrane-associated protein (APMAP) has been described as an adipogenic factor in 3T3-L1 cells with unknown biochemical function; we therefore aimed to investigate the physiologic function of APMAP
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A140
Arecaidine propargyl ester hydrobromide, solid
C10H13NO2 · HBr