EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Effect of fermented and unfermented buckwheat flour on functional properties of gluten-free muffins.
PMID 28559601
Abstract
Selected functional properties of four types of gluten-free muffins made of unfermented and fermented (by
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238813
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
C14H18O4