EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Hepatoprotective activity of chrysin is mediated through TNF-α in chemically-induced acute liver damage: An
PMID 28565752
Abstract
Chrysin (5,7-dihydroxyflavone) is a naturally occurring flavonoid present at high levels in honey, propolis and numerous plant extracts. Chrysin is known to have hepatoprotective activity, however, the mechanisms by which it exerts this effect remain unclear. In the present study, the effects of chrysin in carbon tetrachloride (CCl
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C80105
Chrysin, 97%
C15H10O4
95082
Chrysin, analytical standard
C15H10O4