EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Global methylation and promoter-specific methylation of the
PMID 28565861
Abstract
Tumor suppressor gene promoter CpG island methylation is a well-recognized mechanism in cancer pathogenesis, but its role in multiple myeloma (MM) is controversial. The present study investigated the methylation status and expression of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13438
Sodium bisulfite solution, purum, ~40%
HNaO3S