EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Gender difference in kidney electrolyte transport. I. Role of AT
PMID 28566500
Abstract
We studied gender differences in Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T133
Inactin® hydrate, ≥98% (HPLC)
C10H15N2NaO2S · xH2O