EMAIL THIS PAGE TO A FRIEND
Scientific reports
Mathematical Analysis of Glioma Growth in a Murine Model.
PMID 28566701
Abstract
Five immunocompetent C57BL/6-cBrd/cBrd/Cr (albino C57BL/6) mice were injected with GL261-luc2 cells, a cell line sharing characteristics of human glioblastoma multiforme (GBM). The mice were imaged using magnetic resonance (MR) at five separate time points to characterize growth and development of the tumor. After 25 days, the final tumor volumes of the mice varied from 12 mm