EMAIL THIS PAGE TO A FRIEND
3 Biotech
Analysis of toxic, antidiabetic and antioxidant potential of Bambusa balcooa Roxb. leaf extracts in alloxan-induced diabetic rats.
PMID 28567632
Abstract
Bambusa balcooa (Poaceae) is native to India and has been used traditionally by the tribes of Northeast India to treat diabetes. The present investigation was aimed to evaluate the toxicity, anti-diabetic activity along with in vitro antioxidant activity of the leaf of B. balcooa in alloxan-induced diabetic rats and also identify active compounds by using HPLC. The acute toxicity test of aqueous extract of B. balcooa leaf revealed that the median lethal dose (LD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108383
1,1,3,3-Tetramethoxypropane, 99%
C7H16O4