EMAIL THIS PAGE TO A FRIEND
The Journal of nutritional biochemistry
α-Glucosidase inhibitory effect of resveratrol and piceatannol.
PMID 28570943
Abstract
Dietary polyphenols have been shown to inhibit α-glucosidase, an enzyme target of some antidiabetic drugs. Resveratrol, a polyphenol found in grapes and wine, has been reported to inhibit the activity of yeast α-glucosidase. This triggered our interest to synthesize analogs and determine their effect on mammalian α-glucosidase activity. Using either sucrose or maltose as substrate resveratrol, piceatannol and 3'-hydroxypterostilbene showed strong inhibition of mammalian α-glucosidase activity; pinostilbene, cis-desoxyrhapontigenin and trans-desoxyrhapontigenin had moderate inhibition. Compared to acarbose (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0453
Piceatannol, powder
C14H12O4