EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Quick and sensitive SPR detection of prion disease-associated isoform (PrP
PMID 28570989
Abstract
Herein, we constructed a novel sandwich surface plasmon resonance (SPR) detection assay for sensitive prion disease-associated isoform (PrP