EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Rapid multiresidue determination of bisphenol analogues in soil with on-line derivatization.
PMID 28573319
Abstract
Bisphenol analogues are compounds extensively used which have been potentially linked to adverse health effects. Nevertheless, few studies reported the analysis of compounds, other than bisphenol A, in environmental solid samples and none in soil samples. In this study, a rapid and sensitive analytical method is presented for the simultaneous determination of 13 bisphenols in soil samples. The method combines ultrasonic-assisted extraction of samples placed in small columns and GC-MS/MS analysis. Manual and on-line derivatizations were compared and results showed that significant higher chromatographic responses were achieved with on-line derivatization. Different parameters such as the quantity of derivatization agent, the extraction solvent, or the extraction time were assayed. The detection limits for all target bisphenols ranged from 0.04 to 0.27 ng g
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73081
Tetrachlorobisphenol A, analytical standard
C15H12Cl4O2