EMAIL THIS PAGE TO A FRIEND
Journal of applied biomaterials & functional materials
Effect of storage medium and aging duration on mechanical properties of self-adhesive resin-based cements.
PMID 28574094
Abstract
The aim of this study was to investigate the effects of storage medium and aging duration on Martens hardness (HM) and indentation modulus (EIT) of self-adhesive resin-based cements (SARCs). A total of 416 discs were fabricated from 8 SARCs (n = 52 per SARC): (i) BeautyCem (BEA), (ii) Bifix SE (BIF), (iii) Clearfil SA Cement Automix (CLE), (iv) RelyX Unicem2 Automix (RXU), (v) SeT (SET), (vi) SmartCem 2 (SMC), (vii) SoloCem (SOC) and (viii) SpeedCEM (SPC). The specimens were ground and stored in (a) physiological saliva, (b) artificial saliva, (c) sodium chloride and (d) distilled water, at 37°C for 1, 7, 14, 28, 90 and 180 days. Non-aged specimens (3 hours after photo-initiation) of each SARC acted as control groups. HM and EIT were assessed using a universal hardness testing machine. Data were analyzed using Kolmogorov-Smirnov and Kruskal-Wallis tests and paired t-test, and 3-, 2- and 1-way ANOVA with post hoc Scheffé test (p<0.05). SARC materials exerted the highest influence on HM and EIT values (p<0.001, partial eta squared [ηP²] = 0.753 and 0.433, respectively), closely followed by aging duration (p<0.001, ηP² = 0.516 and 0.255) and storage medium (p<0.001, ηP² = 0.043 and 0.033). The interaction effect of the combinations of the 3 independent parameters was also significant (p<0.001, ηP² = 0.163 and 0.133). The lowest initial HM and EIT was presented by CLE, followed by SET and SPC; highest HM was for BIF, followed by RXU and SMC. SET gave a greater HM and EIT percentage decrease than SOC, SPC, BEA, SMC, RXU or CLE. Aging duration is a major factor affecting the micromechanical properties of SARCs, while storage medium was shown to have a significant but minor role.