EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Synthesis of xanthan gum graft copolymer and its application for controlled release of highly water soluble Levofloxacin drug in aqueous medium.
PMID 28578956
Abstract
Graft copolymers (XG-g-PNVP-1 to XG-g-PNVP-5) of xanthan gum (XG) and poly(N-vinyl-2-pyrrolidone) (PNVP) was synthesized by free radical polymerization using peroxymonosulphate/thiourea redox pair. The synthesized graft copolymers were well characterized by