EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
Cyanogenic Glucosides and Derivatives in Almond and Sweet Cherry Flower Buds from Dormancy to Flowering.
PMID 28579996
Abstract
Almond and sweet cherry are two economically important species of the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68264
α-Hydroxyisobutyronitrile β-D-glucopyranoside, ≥98.0% (HPLC)
C10H17NO6