Multiple Disease Resistance Loci Affect Soilborne Disease Resistance in Tobacco (Nicotiana tabacum).
PMID 28581342
Phytophthora nicotianae and Ralstonia solanacearum are two of the most important pathogens affecting tobacco worldwide. Greater insight regarding genetic systems controlling resistance to these two soilborne pathogens, as well as identification of DNA markers associated with genomic regions controlling this resistance, could aid in variety development. An evaluation of 50 historical tobacco lines revealed a high positive correlation between resistances to the two pathogens, preliminarily suggesting that some genomic regions may confer resistance to both pathogens. A quantitative trait loci (QTL) mapping experiment designed to investigate the genetic control of soilborne disease resistance of highly resistant 'K346' tobacco identified four QTL significantly associated with resistance to P. nicotianae (explaining 60.0% of the observed phenotypic variation) and three QTL to be associated with R. solanacearum resistance (explaining 50.3% of the observed variation). The two QTL with the largest effect on Phytophthora resistance were also found to be the QTL with the greatest effects on resistance to Ralstonia. This finding partially explains previously observed associations between resistances to these two pathogens among U.S. current cultivars and within breeding populations. Further study is needed to determine whether these relationships are due to the same genes (i.e., pleiotropy) or favorable coupling-phase linkages that have been established over time.