EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Improved respirable fraction of budesonide powder for dry powder inhaler formulations produced by advanced supercritical CO
PMID 28583335
Abstract
A budesonide (BDS) suspension was obtained via advanced supercritical carbon dioxide (scCO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388920
Sorbitan monopalmitate
C22H42O6