EMAIL THIS PAGE TO A FRIEND
Antiviral research
Ubiquitin C-terminal hydrolase-L3 promotes interferon antiviral activity by stabilizing type I-interferon receptor.
PMID 28583475
Abstract
Type-I interferons (IFN-I) are important antiviral drugs which are widely used in clinical therapy of diverse viral infections. However, understanding the detailed mechanisms for IFN-I antiviral signaling remains a major challenge, and may provide novel targets for IFN-based antiviral therapy. So far, the roles of deubiquitinases (DUBs) in regulating IFN-I antiviral activity are still largely unexplored. Here, we find that Ubiquitin C-terminal hydrolase-L3 (UCHL3) plays an important role in regulating type I-interferon (IFN-I) mediated antiviral response. Interestingly, we find that UCHL3 regulates COPS5-dependent deneddylation of Cullin1, which is an essential component of SCF