EMAIL THIS PAGE TO A FRIEND
Hypertension (Dallas, Tex. : 1979)
NEFM (Neurofilament Medium) Polypeptide, a Marker for Zona Glomerulosa Cells in Human Adrenal, Inhibits D1R (Dopamine D1 Receptor)-Mediated Secretion of Aldosterone.
PMID 28584012
Abstract
Heterogeneity among aldosterone-producing adenomas (APAs) has been highlighted by the discovery of somatic mutations.