EMAIL THIS PAGE TO A FRIEND
Molecular psychiatry
Rescue of impaired sociability and anxiety-like behavior in adult cacna1c-deficient mice by pharmacologically targeting eIF2α.
PMID 28584287
Abstract
CACNA1C, encoding the Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0843
ISRIB, ≥98% (HPLC)
C22H24Cl2N2O4