EMAIL THIS PAGE TO A FRIEND
Journal of clinical biochemistry and nutrition
Compound 48/80, a mast cell degranulator, causes oxidative damage by enhancing vitamin C synthesis via reduced glutathione depletion and lipid peroxidation through neutrophil infiltration in rat livers.
PMID 28584400
Abstract
In this study, we examined whether compound 48/80 (C48/80), a mast cell degranulator, causes hepatic oxidative damage in rats. Serum and liver biochemical parameters were determined 0.5, 3 or 6 h after a single treatment with C48/80 (0.75 mg/kg). Serum histamine and serotonin levels increased 0.5 h after C48/80 treatment but diminished thereafter. Increases in serum vitamin C (VC) and transaminases and hepatic hydrogen peroxide, lipid peroxide, and myeloperoxidase levels and a decrease in hepatic reduced glutathione level occurred 0.5 h after C48/80 treatment and further proceeded at 3 h, but these changes diminished at 6 h. Serum lipid peroxide and hepatic VC levels increased 3 h after C48/80 treatment. Hepatic glycogen level decreased 0.5 h after C48/80 treatment and further decreased at 3 h. Pre-administered ketotifen diminished all these changes found at 3 h after treatment, while pre-administered NPC 14686 diminished these changes except changes in serum histamine and serotonin levels. Hepatocellular apoptosis observed at 3 h after C48/80 treatment was attenuated by pre-administered ketotifen and NPC 14686. These results indicate that C48/80 causes oxidative damage by enhancing VC synthesis via reduced glutathione depletion-dependent glycogenolysis and lipid peroxidation through neutrophil infiltration following mast cell degranulation in rat livers.