EMAIL THIS PAGE TO A FRIEND
PeerJ
Cucurbitacins: elucidation of their interactions with the cytoskeleton.
PMID 28584704
Abstract
Cucurbitacins, a class of toxic tetracyclic triterpenoids in Cucurbitaceae, modulate many molecular targets. Here we investigated the interactions of cucurbitacin B, E and I with cytoskeletal proteins such as microtubule and actin filaments. The effects of cucurbitacin B, E and I on microtubules and actin filaments were studied in living cells (Hela and U2OS) and
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0577
Cucurbitacin E, ≥95% (HPLC)
C32H44O8