EMAIL THIS PAGE TO A FRIEND
AMB Express
Expanding the range of polyhydroxyalkanoates synthesized by methanotrophic bacteria through the utilization of omega-hydroxyalkanoate co-substrates.
PMID 28587442
Abstract
The first methanotrophic syntheses of polyhydroxyalkanoates (PHAs) that contain repeating units beyond 3-hydroxybutyrate and 3-hydroxyvalerate are reported. New PHAs synthesized by methanotrophs include poly(3-hydroxybutyrate-co-4-hydroxybutyrate) (P(3HB-co-4HB)), poly(3-hydroxybutyrate-co-5-hydroxyvalerate-co-3-hydroxyvalerate) (P(3HB-co-5HV-co-3HV)), and poly(3-hydroxybutyrate-co-6-hydroxyhexanoate-co-4-hydroxybutyrate) (P(3HB-co-6HHx-co-4HB)). This was achieved from a pure culture of Methylocystis parvus OBBP where the primary substrate is methane and the corresponding ω-hydroxyalkanoate monomers are added as a co-substrate after the cells are subjected to nitrogen-limited conditions.