EMAIL THIS PAGE TO A FRIEND
ACS nano
Injectable Fullerenol/Alginate Hydrogel for Suppression of Oxidative Stress Damage in Brown Adipose-Derived Stem Cells and Cardiac Repair.
PMID 28590722
Abstract
Stem cell implantation strategy has exhibited potential to treat the myocardial infarction (MI), however, the low retention and survival limit their applications due to the reactive oxygen species (ROS) microenvironment after MI. In this study, the fullerenol nanoparticles are introduced into alginate hydrogel to create an injectable cell delivery vehicle with antioxidant activity. Results suggest that the prepared hydrogels exhibit excellent injectable and mechanical strength. In addition, the fullerenol/alginate hydrogel can effectively scavenge the superoxide anion and hydroxyl radicals. Based on these results, the biological behaviors of brown adipose-derived stem cells (BADSCs) seeded in fullerenol/alginate hydrogel were investigated in the presence of H