EMAIL THIS PAGE TO A FRIEND
Journal of microencapsulation
Thermo-responsive magnetic liposomes for hyperthermia-triggered local drug delivery.
PMID 28590788
Abstract
We prepared and characterised thermo-responsive magnetic liposomes, which were designed to combine features of magnetic targeting and thermo-responsive control release for hyperthermia-triggered local drug delivery. The particle size and zeta-potential of the thermo-responsive magnetic ammonium bicarbonate (MagABC) liposomes were about 210 nm and -14 mV, respectively. The MagABC liposomes showed encapsulation efficiencies of about 15% and 82% for magnetic nanoparticles (mean crystallite size 12 nm) and doxorubicin (DOX), respectively. The morphology of the MagABC liposomes was visualised using transmission electron microscope (TEM). The MagABC liposomes showed desired thermo-responsive release. The MagABC liposomes, when physically targeted to tumour cells in culture by a permanent magnetic field yielded a substantial increase in intracellular accumulation of DOX as compared to non-magnetic ammonium bicarbonate (ABC) liposomes. This resulted in a parallel increase in cytotoxicity for DOX loaded MagABC liposomes over DOX loaded ABC liposomes in tumour cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3531
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
C41H82NO8P