EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Rapid gas chromatographic assay for monitoring valnoctamide in plasma.
PMID 2859299
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V4765
Valnoctamide, ≥98% (NMR)
C8H17NO