Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
An in vitro study of the long non-coding RNA TUG1 in tongue squamous cell carcinoma.
PMID 28597931
This study sought to study the expression of the long non-coding RNA (lncRNA) taurine-upregulated gene 1 (TUG1) in tongue squamous cell carcinoma (TSCC) and reveal its possible function. qRT-PCR was used to evaluate 27 samples of fresh TSCC tissues and adjacent normal tongue tissues. siRNA technology was employed to downregulate TUG1 expression in CAL-27 and SCC-9 cell lines. The 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2-H-tetrazolium bromide (MTT) assay was utilized to assess cell proliferation ability; apoptosis and cell-cycle phases were analysed via flow cytometry. qRT-PCR findings indicated that the lncRNA TUG1 was upregulated in TSCC tissues compared with adjacent normal tongue tissues (P<.05). After TUG1 expression was downregulated using siRNA technology, cell proliferation was significantly inhibited (P<.05), and the number of cells in S phase was reduced (P<.05). The lncRNA TUG1 may represent a potential oncogene in TSCC.
Related Materials
Product #
Molecular Formula
Add to Cart
EHU130891 MISSION® esiRNA, esiRNA human TUG1 (esiRNA1)