EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Protective effects of Oviductus Ranae-containing serum on oxidative stress-induced apoptosis in rat ovarian granulosa cells.
PMID 28602868
Abstract
Oviductus Ranae (OR) is a traditional Chinese medicine derived from Rana temporaria chensinensis David, and is known to have a wide variety of pharmacological effects. To investigate the function and mechanism of OR-containing serum in protecting rat ovarian granulosa cells from hydrogen peroxide (H H The results revealed that treatment with OR-containing serum reduced apoptosis and mitochondrial membrane damage in H OR-containing serum protected rat ovarian granulosa cells against H