EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance (San Diego, Calif. : 1997)
Improvements, extensions, and practical aspects of rapid ASAP-HSQC and ALSOFAST-HSQC pulse sequences for studying small molecules at natural abundance.
PMID 28603039
Abstract
Previously we introduced two novel NMR experiments for small molecules, the so-called ASAP-HSQC and ALSOFAST-HSQC (Schulze-Sünninghausen et al., 2014), which allow the detection of heteronuclear one-bond correlations in less than 30s at natural abundance. We propose an improved symmetrized pulse scheme of the basic experiment to minimize artifact intensities and the combination with non-uniform sampling to enable the acquisition of conventional HSQC spectra in as short as a couple of seconds and extremely