The Journal of veterinary medical science
Effectiveness of the liver micronucleus assay using juvenile mice.
PMID 28603212
This study investigated the effectiveness of the liver micronucleus (MN) assay using juvenile mice. Therefore, we analyzed various hepatic cytochrome P450 (CYP)- mediated activities of ethoxyresorufin O-deethylation, pentoxyresorufin O-dealkylation, tolbutamide hydroxylation, bufuralol 1'-hydroxylation, aniline hydroxylation and midazolam 4-hydroxylation by CYP1A, CYP2B, CYP2C, CYP2D, CYP2E and CYP3A, respectively, in non-treated male ICR mice aged between 3 and 8 weeks. The enzyme efficiency levels in 3- and 4-week-old mice were approximately similar to or higher than those in 8-week-old mice, except for CYP1A and CYP2E in 3- and 4-week-old mice, respectively. Since these results suggest that juvenile mice have sufficient activities for most CYP enzymes, we also conducted a liver MN assay using diethylnitrosamine (DEN), a rodent hepatocarcinogen, on male ICR mice aged between 3 and 6 weeks. A peripheral blood (PB) MN assay was performed simultaneously in 4-week-old mice. Assays incorporating DEN produced positive results in 3- and 4-week-old mice and showed a dose-dependent increase in the micronucleated hepatocyte frequencies at 4 weeks. Both the liver MN assay in 5- and 6-week-old mice and the PB MN assay had negative results when using DEN. These results suggest that 3- and 4-week-old mice have micronuclei-inducing potential in the liver to detect genotoxic compounds using the liver MN assay.
Related Materials
Product #
Molecular Formula
Add to Cart
Resorufin pentyl ether