EMAIL THIS PAGE TO A FRIEND
Water, air, and soil pollution
Effect of Biochar Amendment and Ageing on Adsorption and Degradation of Two Herbicides.
PMID 28603318
Abstract
Biochar amendment can alter soil properties, for instance, the ability to adsorb and degrade different chemicals. However, ageing of the biochar, due to processes occurring in the soil over time, can influence such biochar-mediated effects. This study examined how biochar affected adsorption and degradation of two herbicides, glyphosate (N-(phosphonomethyl)-glycine) and diuron (3-(3,4-dichlorophenyl)-1,1-dimethylurea) in soil and how these effects were modulated by ageing of the biochar. One sandy and one clayey soil that had been freshly amended with a wood-based biochar (0, 1, 10, 20 and 30%
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261394
1,1-Dimethylurea, 99%
C3H8N2O