EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
A rare castration-resistant progenitor cell population is highly enriched in Pten-null prostate tumours.
PMID 28603917
Abstract
Castration-resistant prostate cancer is a lethal disease. The cell type(s) that survive androgen deprivation remain poorly described, despite global efforts to understand the various mechanisms of therapy resistance. We recently identified in wild-type (WT) mouse prostates a rare population of luminal progenitor cells that we called LSC