EMAIL THIS PAGE TO A FRIEND
Nanoscale
Breaking the electrical barrier between copper and carbon nanotubes.
PMID 28604888
Abstract
Improving the interface between copper and carbon nanotubes (CNTs) offers a straightforward strategy for the effective manufacturing and utilisation of Cu-CNT composite material that could be used in various industries including microelectronics, aerospace and transportation. Motivated by a combination of structural and electrical measurements on Cu-M-CNT bimetal systems (M = Ni, Cr) we show, using first principles calculations, that the conductance of this composite can exceed that of a pure Cu-CNT system and that the current density can even reach 10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F408
Ferrocene, 98%
C10H10Fe