EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
Synergistic Activity with NOTCH Inhibition and Androgen Ablation in ERG-Positive Prostate Cancer Cells.
PMID 28607007
Abstract
The oncogenic activation of the ETS-related gene (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU076241 MISSION® esiRNA, esiRNA human ERG (esiRNA1)