EMAIL THIS PAGE TO A FRIEND
Bone research
Differential involvement of Wnt signaling in Bmp regulation of cancellous versus periosteal bone growth.
PMID 28607813
Abstract
Bone morphogenetic proteins (Bmp) are well-known to induce bone formation following chondrogenesis, but the direct role of Bmp signaling in the osteoblast lineage is not completely understood. We have recently shown that deletion of the receptor Bmpr1a in the osteoblast lineage with