EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Using Voltammetry to Measure the Relative Hydrogen-Bonding Strengths of Pyridine and Its Derivatives in Acetonitrile.
PMID 28608640
Abstract
The voltammetric behavior of 2,3,5,6-tetramethyl-1,4-phenylenediamine was found to be able to differentiate the hydrogen acceptor abilities of electroinactive pyridine compounds in acetonitrile. Weak and strong hydrogen acceptors were distinguished through the onset of a third oxidation process that came about at sub-stoichiometric amounts of strong hydrogen acceptors, but not in the presence of weak hydrogen acceptors. This additional oxidation reaction occurred at a potential between the two 1 e