EMAIL THIS PAGE TO A FRIEND
Faraday discussions
Facile synthesis of an ultra-stable metal-organic framework with excellent acid and base resistance.
PMID 28608885
Abstract
A novel ultra-stable metal-organic framework, MCIF-1, [Cu
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205540
Copper(I) iodide, 98%
CuI