EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Selective and Fast Adsorption of Perfluorooctanesulfonate from Wastewater by Magnetic Fluorinated Vermiculite.
PMID 28614945
Abstract
A novel magnetic fluorinated adsorbent with selective and fast adsorption of perfluorooctanesulfonate (PFOS) was synthesized via a simple ball milling of Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
164194
Heptafluorobutyric acid, 98%
C4HF7O2
171468
Perfluorooctanoic acid, 95%
C8HF15O2