EMAIL THIS PAGE TO A FRIEND
Veterinary immunology and immunopathology
17β-Estradiol and progesterone decrease MDP induced NOD2 expression in bovine mammary epithelial cells.
PMID 28615128
Abstract
During the periparturient period, many neuroendocrine changes develop in cows. Periparturient hormone fluxes may adversely affect mammary gland immunity and mastitis susceptibility. 17β-Estradiol (E