EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
Increased vitamin D intake differentiated according to skin color is needed to meet requirements in young Swedish children during winter: a double-blind randomized clinical trial.
PMID 28615261