EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
HiHiMap: single-cell quantitation of histones and histone posttranslational modifications across the cell cycle by high-throughput imaging.
PMID 28615324
Abstract
We describe