EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel regulatory mechanism of serine biosynthesis associated with 3-phosphoglycerate dehydrogenase in Arabidopsis thaliana.
PMID 28615699
Abstract
The proteinogenic amino acid L-serine is a precursor for various essential biomolecules in all organisms. 3-Phosphoglycerate dehydrogenase (PGDH) is the first committed enzyme of the phosphorylated pathway of L-serine biosynthesis, and is regulated by negative feedback from L-serine in bacteria and plants. In the present study, two Arabidopsis PGDH isoforms were inhibited by L-serine but were activated by L-amino acids such as L-homocysteine in vitro. Activation and inhibition by these amino acids was cooperative, suggesting an allosteric mechanism. Moreover, the half maximal effective concentration of L-homocysteine was 2 orders of magnitude lower than that of L-serine, suggesting greater regulatory potency. These are the first data to show that PGDH is activated by various biomolecules and indicate that serine biosynthesis is regulated by multiple pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69453
L-Homocysteine, ≥98.0% (NT)
C4H9NO2S