EMAIL THIS PAGE TO A FRIEND
Evolutionary applications
Human management and hybridization shape treegourd fruits in the Brazilian Amazon Basin.
PMID 28616065
Abstract
Local people's perceptions of cultivated and wild agrobiodiversity, as well as their management of hybridization are still understudied in Amazonia. Here we analyze domesticated treegourd (