EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Antimosquito Activity of a Titanium-Organic Framework Supported on Fabrics.
PMID 28618230
Abstract
Waste swamps, stagnant water, and poor hygiene practices result in the proliferation of mosquitoes that may cause transmissible and infectious diseases such as malaria, typhoid, cholera, and Zika virus sickness. It has been shown that composites of the traditional natural fibers cotton, viscose, and linen and a Ti-bearing metal-organic framework, NH
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
381071
2-Aminoterephthalic acid, 99%
C8H7NO4